Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Neuronal Differentiation Inducer III

Catalog No. 48074310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-074-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-074-310MG Supplier MilliporeSigma™ Supplier No. 48074310MG
Add to Cart
Add to Cart

A cell-permeable isoxazole compound that selectively induces robust neuronal differentiation (Effective concentration = 20 μM) in various stem/progenitor cells.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 937807-66-4
Molecular Formula C11H10N2O2S
Formula Weight 234.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.