Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Inhibitor IV

Catalog No. 1240111MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-401-11MG 1 mg
12-401-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-401-11MG Supplier MilliporeSigma™ Supplier No. 1240111MG
Add to Cart
Add to Cart

The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins Akt
CAS 681281-88-9
Molecular Formula C31H27IN4S
Formula Weight 614.6g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.