Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NF-κB Activation Inhibitor VII, CID-2858522

Catalog No. 48045710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-045-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-045-710MG Supplier MilliporeSigma™ Supplier No. 48045710MG
Add to Cart
Add to Cart

The NF-κB Activation Inhibitor VII, CID-2858522 controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

A cell-permeable selective inhibitor of NF-κB activation induced by PKC pathway activators such as PMA (IC50 = 70 nM) in a HEK293 cell-based NF-κB luciferase reporter gene assay, by operating downstream of PKC but upstream of IKKβ, without inhibiting other NF-κB activation pathways. At 10μM, this compound only inhibits Raf, TLK1, and JAK2 by 57%, 70%, and 53%, respectively, among a panel of 353 protein kinases. In addition, it attenuates CD3/CD28 and PMA/ionomycin-induced production of IL-2 by Jurkat T-cells (IC50 < 5μM) and anti-IgM-stimulated proliferation of murine B-lymphocytes (IC50 = 2μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description >95% by HPLC
Molecular Formula C28H39N3O3
Formula Weight 465.63g/mol
Solubility DMSO
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.