Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NF023

Catalog No. 48041510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-041-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-041-510MG Supplier MilliporeSigma™ Supplier No. 48041510MG
Add to Cart
Add to Cart

A suramin analog that acts as a selective and direct G-protein antagonist for α-subunits of the Go

A suramin analog that acts as a selective and direct G-protein antagonist for α-subunits of the Go/Gi group (EC50 ∽ 300nM). Suppresses GTPγS binding to recombinant G-protein α-subunits. Acts by inhibiting the formation of the agonist-specific ternary complex (agonist/receptor/G-protein). NF023 does not interfere with the interaction between α-subunits and βγ-dimer but competes with the effector binding site. Also a P2X receptor antagonist (IC50 = 240nM for P2X1; 8.5μM for P2X3 receptor in rat muscle). Causes a concentration dependent inhibition of excitatory junction potentials (IC50 = 4.8 pM).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins α-subunits of the Go/Gi group of G-protein
Molecular Formula C35H20N4O21S6·6Na
Formula Weight 1162.9g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.