Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NFAT Inhibitor

Catalog No. 480402500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-040-2500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-040-2500UG Supplier MilliporeSigma™ Supplier No. 480402500UG
Add to Cart
Add to Cart

The NFAT Inhibitor controls the biological activity of NFAT. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Met-Ala-Gly-Pro-His-Pro-Val-Ile-Val-Ile-Thr-Gly-Pro-His-Glu-Glu-OH
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins NFAT (Nuclear Factor of Activated T-cells)
Molecular Formula C75H118N20O22S
Formula Weight 1683.9g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.