Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NFAT Inhibitor, MCV1

Catalog No. 4804042MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-040-42MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-040-42MG Supplier MilliporeSigma™ Supplier No. 4804042MG
Add to Cart
Add to Cart

The NFAT Inhibitor, MCV1 controls the biological activity of NFAT.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Molecular Formula C51H75N10O12
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.