Learn More
MilliporeSigma™ Calbiochem™ NG,NG'-Dimethyl-L-arginine, Dihydrochloride
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Color | White |
| Description | 98% by TLC |
| Target Proteins | NOS in vitro |
| Molecular Formula | C8H18N4O2·2HCl |
| Formula Weight | 275.2g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Quantity | 25 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.