Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nicotinamide Phosphoribosyltransferase Inhibitor, FK866

Catalog No. 4819082MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-190-82MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-190-82MG Supplier MilliporeSigma™ Supplier No. 4819082MG
Add to Cart
Add to Cart

The Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, also referenced under CAS 658084-64-1, controls the biological activity of Nicotinamide Phosphoribosyltransferase. This small molecule/inhibitor is primarily used for Neuroscience applications.

A cell-permeable pyridinylacrylamide compound that acts as a selective, allosteric NAPRT/NAMPT (nicotinamide phosphoribosyltransferase) inhibitor (Ki = 0.4nM for the enzyme/substrate complex; Ki = 0.3nM for the free enzyme), while exhibiting no effect toward NPRT (nicotinic acid phosphoribosyltransferase) activity (82% inhibition of NAPRT activity at 10nM vs no inhibition of NPRT activity at 1μM in K-562 extract). FK866 treatment is shown to induce cell death by depleting NAD+ (Cat. Nos. 481911 and 481915) in HepG2 and NIH-3T3 cultures (by >95% after 24 h treatment of 10nM FK866), likewise the addition of exogenous NAD+ is demonstrated to rescue NIH-3T3 and SH-SY5Y from FK866-induced NAD+-depletion and cell death.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Formulation Supplied as a 50 mM (2mg/102.17μl) solution in DMSO.
Description 97% by HPLC
CAS 658084-64-1
Molecular Formula C24H29N3O2
Formula Weight 391.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.