Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nicotinamide

Catalog No. 481907100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Nicotinamide; Qty: 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
48-190-7100GM Nicotinamide; Qty: 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-190-7100GM Supplier MilliporeSigma™ Supplier No. 481907100GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acts as precursor of coenzymes NAD+ and NADP+

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive Nicotinamide
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.