Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nicotinic Acid

Catalog No. 481918100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Nicotinic Acid; Qty: 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
48-191-8100GM Nicotinic Acid; Qty: 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-191-8100GM Supplier MilliporeSigma™ Supplier No. 481918100GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Component of coenzymes NAD+ and NADP+

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Additive Nicotinic Acid
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.