Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nifuroxazide

Catalog No. 481984500MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-198-4500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-198-4500MG Supplier MilliporeSigma™ Supplier No. 481984500MG
Add to Cart
Add to Cart

A cell-permeable and orally available nitrofuran-based antidiarrheal agent that effectively suppresses the activation of cellular STAT1/3/5 transcription activity.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
Description 97% by HPLC
CAS 965-52-6
Molecular Formula C12H9N3O5
Formula Weight 275.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.