Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nitric Oxide Synthase, Neuronal Inhibitor I

Catalog No. 4900705MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-007-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-007-05MG Supplier MilliporeSigma™ Supplier No. 4900705MG
Add to Cart
Add to Cart

The Nitric Oxide Synthase, Neuronal Inhibitor I, also referenced under CAS 357965-99-2, controls the biological activity of neuronal Nitric Oxide Synthase. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

A cell-permeable, potent and highly selective inhibitor of neuronal nitric oxide synthase (nNOS); (Ki = 120nM). Displays >2500-fold and 320-fold selectivity over eNOS and iNOS, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Formulation Supplied as a dihydrate.
Description 95% by HPLC
Target Proteins nNOS
CAS 357965-99-2
Molecular Formula C8H21N7O2·3TFA·2H2O
Formula Weight 625.4g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.