Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NMDAR2C/2D Inhibitor, DQP-1105

Catalog No. 45458610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-458-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-458-610MG Supplier MilliporeSigma™ Supplier No. 45458610MG
Add to Cart
Add to Cart

The NMDAR2C/2D Inhibitor, DQP-1105 controls the biological activity of NMDAR2C/2D. This small molecule/inhibitor is primarily used for Neuroscience applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow-white
Description 97% by HPLC
Molecular Formula C29H24BrN3O4
Formula Weight 558.42g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.