Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ nor-Binaltorphimine Dihydrochloride

Catalog No. 5080170001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-801-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-801-70001 Supplier MilliporeSigma™ Supplier No. 5080170001
Add to Cart
Add to Cart

A potent antagonist highly selective for κ opioid receptor with no effect onμ- or &delta- opioid receptors

A potent antagonist highly selective for κ opioid receptor with no effect onμ- or &delta- opioid receptors. Widely used in studies related to pain, addition, anxiety, depression, and sleep.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins kappa opioid receptor
Molecular Formula C40H43N3O6·2HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.