Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Normal Guinea Pig Serum, Lyophilized Solid

Catalog No. 5664005ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-640-05ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-640-05ML Supplier MilliporeSigma™ Supplier No. 5664005ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use in radioimmunoassay. MilliporeSigma™; Calbiochem™ Normal Guinea Pig Serum, Lyophilized Solid is normal carrier sera, produced from pooled lots of sera from healthy, non-immunized guinea pigs.

  • Form: White lyophilized solid
  • Formulation: Lyophilized from 0.25M NaCl, 0.01M Sodium Phosphate
  • Preservative: ≤0.1% sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Ok to freeze. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Species Non-immunized guinea pig
Form Lyophilized
Product Type Guinea Pig Serum
Quantity 5 mL
Formulation Lyophilized from 0.25M NaCl, 0.01M Sodium Phosphate
For Use With (Application) Cell culture
Lyophilized Yes
pH 7.6
Preservative ≤0.1% sodium azide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.