Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Notch Transcription Factor Inhibitor, SAHM1

Catalog No. 4910021MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-100-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-100-21MG Supplier MilliporeSigma™ Supplier No. 4910021MG
Add to Cart
Add to Cart

Notch Transcription Factor Inhibitor, SAHM1, is a cell-permeable inhibitor that prevents assembly in the NOTCH1 trans-activation complex.

Peptide Sequence: Acetyl-βAla-ERLRRRI*LCR*HHST* = Two S5 molecules (a non-natural alkenyl amino acid) stapled together with a double bond
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 93% by HPLC
Molecular Formula C94H162N36O23S·7CF3CO2H·13H2O
Formula Weight 3228.9g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.