Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NOX Inhibitor VII, Thr101

Catalog No. 5005260001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-052-60001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-052-60001 Supplier MilliporeSigma™ Supplier No. 5005260001
Add to Cart
Add to Cart

A cell-permeable, selenium-to-sulfur substituted Ebselen (Cat

A cell-permeable, selenium-to-sulfur substituted Ebselen (Cat. No. 324483) derivative that exhibits similar NOX2 inhibitory activity as Ebselen by blocking p22 phox c-terminal PRD (proline-rich domain) and p47 phox bis -SH3 interaction. Although less potent than Ebselen in cell-free p22 phox PRD-p47 phox bis -SH3 binding (IC50 = 4 vs. 0.3μM) and NOX2 activity (IC50 = 4 vs. 0.6μM) assays, Thr101 is more potent and NOX2-selective than Ebselen in cell-based assays (Thr101 IC50 /Ebselen IC50 = 0.3/0.5, 3/0.15, 8/no inhibition, 8/0.7μM, respectively, against NOX2, NOX1, NOX4, and NOX5) and exhibits no glutathione peroxidase activity.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Description 98% by HPLC
Target Proteins blocks p22phox c-terminal PRD (proline-rich domain) and p47phox bis-SH3 interaction
Molecular Formula C14H10FNOS
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.