Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NOX1 Inhibitor, ML171

Catalog No. 49200210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-200-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-200-210MG Supplier MilliporeSigma™ Supplier No. 49200210MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NOX1 Inhibitor, ML171, also referenced under CAS 6631-94-3, controls the biological activity of NOX1.

A highly selective, cell-permeable, and reversible 2-acetylphenothiazine that is shown to inhibit NADPH Oxidase-1 (Nox1) (IC50 = 0.129μM, and 0.25μM) in human HT29 and HEK293 cell-based assays, respectively. Unlike most other currently used Nox inhibitors, it only has marginal activity on other cellular ROS-producing enzymes and receptors including the other Nox isoforms (IC50 = 5μM, 3μM and 5μM for Nox2, Nox3, and Nox4, respectively) and xanthine oxidase, XO, (IC50 = 5.5μM) in HEK293 cell-based assays. It is also shown to block Nox1-dependent ROS generation in HEK293 cultures dose-dependently at concentrations up to ∽ 100nM; however, its inhibitory effect is reversed through over-expression of Nox1, thereby suggesting that this compound is highly selective for Nox1, with little effect against Nox2-dependent ROS generation (IC50 > 10μM). Qualitatively, it displays an inhibitory effect against SrcYF-induced ECM-degrading invadopodia formation in DLD1 Human Colon Cancer Cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Yellow-orange
CAS 6631-94-3
Molecular Formula C14H11NOS
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.