Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nrf2 Activator III, TAT-14 Peptide

Catalog No. 49204210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-204-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-204-210MG Supplier MilliporeSigma™ Supplier No. 49204210MG
Add to Cart
Add to Cart

The Nrf2 Activator III, TAT-14 Peptide modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Biochemicals applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg∼Leu-Gln-Leu-Asp-Glu-Glu-Thr-Gly-Glu-Phe-Leu-Pro-Ile-Gln-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Molecular Formula C137H230N48O39
Formula Weight 3173.6g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.