Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NS-398

Catalog No. 3492545MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-925-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-925-45MG Supplier MilliporeSigma™ Supplier No. 3492545MG
Add to Cart
Add to Cart

A cell-permeable selective inhibitor of COX-2 in vitro.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by TLC
Target Proteins COX-2
CAS 123653-11-2
Molecular Formula C13H18N2O5S
Formula Weight 314.4g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.