Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nuclease (Staphylococcus aureus)

Catalog No. 49289915KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-289-915KU 15 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-289-915KU Supplier MilliporeSigma™ Supplier No. 49289915KU

missing translation for 'validMainframeMsg'

Cleaves RNA DNA to produce oligo- mononucleotides.

EMD Millipore Calbiochem™ Nuclease (Staphylococcus aureus) cleaves DNA RNA with terminal 3'-phosphate groups.

  • Native Staphylococcus aureus nuclease
  • Converts stared rabbit reticulocyte cell-free extracts (lysate) into mRNA-dependent protein synthesis systems
  • Note: 1 KU = 1000 units
  • Packaged in glass bottle, hygroscopic, lyophilized solid
  • CAS: 9013-53-0
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Product Type Cell Lysis Enzyme
Content And Storage 2°C to 8°C
Quantity 15 kU
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.