Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Okadaic Acid, Ammonium Salt

Catalog No. 45961625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-961-625UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-961-625UG Supplier MilliporeSigma™ Supplier No. 45961625UG
Add to Cart
Add to Cart

Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.

Water-soluble analog of Okadaic Acid (Cat. No. 495604). Inhibits protein phosphatases 1 and 2A.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins PP1, PP2A
CAS 155716-06-6
Molecular Formula C44H67O13·NH4
Formula Weight 822g/mol
Solubility DMSO, EtOH, or H2O
Content And Storage Protect from light
Form Solid
Quantity 25 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.