Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Okadaic Acid, Sodium Salt

Catalog No. 45962025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-962-025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-962-025UG Supplier MilliporeSigma™ Supplier No. 45962025UG
Add to Cart
Add to Cart

Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.

Water-soluble analog of Okadaic Acid (Cat. No. 495604). Inhibits protein phosphatases 1 and 2A.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Colorless
Description 98% by TLC
Target Proteins PP1, PP2
Molecular Formula C44H67O13·Na
Formula Weight 827g/mol
Solubility DMSO, EtOH, or H2O
Content And Storage Protect from light
Form Solid to Film
Quantity 25 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.