Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ 100X TE Buffer Solution Liquid Concentrate
Shop All MilliporeSigma ProductsDescription
- Molecular Biology grade: D
- pH of solution at 25°C: 6.85 to 7.15
- Protease: None detected
- RNase: None detected
- Color: White
Specifications
Specifications
| CAS | 7732-18-5 |
| For Use With (Application) | Molecular Biology |
| Physical Form | Fine Powder |
| Quantity | 1 Pk. |
| Solution Type | Buffer |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.