Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ 10X TAE Buffer, Liquid Concentrate

Catalog No. 872520L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-252-0L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 87-252-0L Supplier MilliporeSigma™ Supplier No. 872520L

missing translation for 'validMainframeMsg'

For use in molecular biology applications

  • Molecular Biology grade: D
  • pH of solution at 25°C: 6.85 to 7.15
  • Protease: None detected
  • RNase: None detected
TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.