Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Acrylamide

Catalog No. 11705KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-705-KG 5 kg
11-501-00GM 100 g
11-601-KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-705-KG Supplier MilliporeSigma™ Supplier No. 11705KG

missing translation for 'validMainframeMsg'

Suitable for polyacrylamide gel electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

pH Range 5.5 to 6.5
Form Liquid
Quantity 5 kg
Reagent Type Acrylamide
Storage Requirements 15-30°C

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.