Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Acrylamide: Bis-acrylamide 29:1, 40% Solution

Catalog No. 17101L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-101-L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-L Supplier MilliporeSigma™ Supplier No. 17101L

missing translation for 'validMainframeMsg'

Suitable for nucleic acids and protein separations and sequencing

  • Molecular biology grade
  • Hazard of handling powdered acrylamide is eliminated with 40% solution in water
  • Reconstitute with deionized water to desired final percentage
  • Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety
  • Yields 40% working solution of 29: 1 ratio
  • DNase/Protease/RNase: None detected
TRUSTED_SUSTAINABILITY

Specifications

Form Clear, colorless solution
Quantity 1 L
Reagent Type Acrylamide: Bis-acrylamide
Storage Requirements 2- 8°C

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.