Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Acrylamide: Bis-acrylamide 29:1, Premixed Powder

Catalog No. 1600200GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-002-00GM 200 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-002-00GM Supplier MilliporeSigma™ Supplier No. 1600200GM

missing translation for 'validMainframeMsg'

Suitable for nucleic acids and protein separations and sequencing

  • Molecular biology grade
  • Reconstitute with deionized water to final volume of 500mL for 200 g size
  • Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety
  • Yields 40% working solution of 29: 1 ratio
  • DNase/Protease/RNase: None detected
TRUSTED_SUSTAINABILITY

Specifications

Form White free-flowing powder
Quantity 200 g
Reagent Type Acrylamide: Bis-acrylamide
Storage Requirements 15-30°C

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.