Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Agarose, Molecular Biology Grade, Low Melting

Catalog No. 2070100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-701-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-701-00GM Supplier MilliporeSigma™ Supplier No. 2070100GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for IEF electrophoresis.

EMD Millipore Calbiochem™ OmniPur™ Agarose, Molecular Biology Grade, Low Melting is preparative agarose.

  • Allows for digestions, ligations in melted gel
  • Easy recovery from gel slices
  • Molecular Biology Grade
  • DNase: None Detected
  • RNase: None Detected
  • Protease: None Detected
  • EEO (-mr): 0.15 max
  • Gelling range (1%): 24-29°C
  • Gel Strength (1%): 250g/cm2 min
  • Melting point (1%): 62 to 68°C
  • Sulfate (SO4): ≤0.15%
  • CAS: 9012-36-6
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 100 g
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.