Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Agarose Super-Fine Resolution

Catalog No. 2082250GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-822-50GM 250 g
20-811-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-822-50GM Supplier MilliporeSigma™ Supplier No. 2082250GM

Suitable for microsatellite analysis, RFLP AFLP.

EMD Millipore Calbiochem™ OmniPur™ Agarose Super-Fine Resolution is used for analytical preparative electrophoresis.

  • Provides 2% resolution in 200 bp to 1,000 bp range
  • Specifications:
  • Ash: ≤0.35%
  • Conductivity (1%, water): ≤10μmho
  • DNase: None Detected
  • EEO (-mr): ≤0.12
  • Gelling range (1.5%): ≤30.0°C
  • Gel strength (1.5%): ≥500g/cm
  • Moisture (H2O) ≤7.0%
  • Protease: None Detected
  • RNase: None Detected
  • Sulfate (SO4): ≤0.11%
  • CAS: 9012-36-6
TRUSTED_SUSTAINABILITY

Specifications

Color White
Melting Temperature At 1.5%, ≤70.0°C
Quantity 250 g
Storage Requirements 15° to 30°C
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.