Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ DTT

Catalog No. 387025GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-702-5GM 25 g
38-605-GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-702-5GM Supplier MilliporeSigma™ Supplier No. 387025GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for reduction of protein disulfide linkages.

  • Molecular Biology grade
  • A (0.02M in water): ≤0.05 AU
  • Assay (-SH content): ≥99.4%
  • DNase: None Detected
  • Infrared spectrum: Conforms to standard
  • Loss on drying: ≤0.5%
  • Melting point: 39 to 43°C
  • Protease: None Detected
  • RNase: None Detected
  • Solubility (5%, water): To pass test
  • Packaged in Glass Bottle
  • Assay: ≥99.4%
  • CAS: 3483-12-3
  • Packaging: Glass Bottle
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.