Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Ethidium Bromide

Catalog No. 43405GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-405-GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-405-GM Supplier MilliporeSigma™ Supplier No. 43405GM

missing translation for 'validMainframeMsg'

Suitable for fluorescent staining of nucleic acids.

EMD Millipore Calbiochem™ OmniPur™ Ethidium Bromide facilitates purification of DNA in Cesium Chloride gradient.

  • Fluorescent stain
  • Molecular biology grade
  • Specifications:
  • Assay (dry basis): ≥98.0%
  • DNase: None Detected
  • Loss on drying: ≤8.0%
  • Protease: None Detected
  • RNase: None Detected
  • CAS: 1239-45-8
  • Molecular formula: C21H20N3Br
TRUSTED_SUSTAINABILITY

Specifications

Description Ethidium Bromide
Color Dark Red/Purple
Quantity 5 g
Product Type Fluorescent DNA/RNA Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.