Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Ethidium Bromide, Molecular Biology Grade

Catalog No. 441010ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-101-0ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-101-0ML Supplier MilliporeSigma™ Supplier No. 441010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for visualization of nucleic acids in gels.

EMD Millipore Calbiochem™ OmniPur™ Ethidium Bromide, Molecular Biology Grade is fluorescent stain which facilitates purification of DNA.

  • Molecular Biology grade
  • Specifications
  • DNase: None Detected
  • RNase: None Detected
  • Protease: None Detected

Recommended Storage

Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Description Ethidium Bromide
Color Dark Red/Purple
Form Liquid
Formulation Supplied as a 10mg/ml solution in deionized water.
Quantity 10 mL
Product Type Fluorescent DNA/RNA Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.