Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Guanidine Hydrochloride, Molecular Biology Grade

Catalog No. 50101KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1kg
5kg
10kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-101-KG 1kg
50-201-0KG 10kg
50-305-KG 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-101-KG Supplier MilliporeSigma™ Supplier No. 50101KG
Add to Cart
Add to Cart

Purification of nucleic acids polypeptides from cell lysates.

EMD Millipore Calbiochem™ OmniPur™ Guanidine Hydrochloride, Molecular Biology Grade is denaturant which inhibits RNase protease activity.

  • Specifications:
  • A260 (0.1M, water): ≤0.05 AU
  • A280 (0.1M, water): ≤0.01 AU
  • DNase: None Detected
  • Heavy metals (as Pb): ≤0.001%
  • Phosphatase: None Detected
  • Protease: None Detected
  • RNase: None Detected
  • Assay: ≥99.5%
  • CAS: 50-01-1
  • Molecular Formula: H2NC(NH)NH2·HCl
  • Color: White
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1kg
Product Type Guanidine Hydrochloride
Content And Storage 15° to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.