Learn More
Description
- Molecular biology grade
- Specifications:
- pKa at 25°C: 5.50-6.50
- Purity: ≥99.0%
- Packaged in Glass Bottle
- Assay: ≥99.0%
- CAS: 145224-94-8
- Molecular Formula: (C4H8ON)CH2CH2SO3H·H2O
- Packaging: Fibre Carton
- Color: White
Specifications
Specifications
| Chemical Name or Material | MES Monohydrate |
| CAS | 145224-94-8 |
| For Use With (Application) | Molecular Biology |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Solution Type | Buffer |
| Molecular Weight (g/mol) | 213.25 |
| Percent Purity | ≥99.0% |
| Grade | OmniPur |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.