Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ PBS 10X Liquid Concentrate, Molecular Biology Grade

Catalog No. 65054L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-054-L 4 L
65-042-0L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-054-L Supplier MilliporeSigma™ Supplier No. 65054L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Makes 10L of 1X PBS Buffer from each liter.

  • Molecular Biology Grade
  • DNase: None Detected
  • pH (10%, water) at 25°C: 7.3 to 7.5
  • RNase: None Detected
  • Sterility: Passes test
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Quantity 4 L
Grade OmniPur, Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.