Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ PBS 10X Liquid Concentrate, Molecular Biology Grade, Sterile
Shop All MilliporeSigma ProductsDescription
- Molecular Biology Grade
- DNase: None Detected
- pH (10%, water) at 25°C: 7.3 to 7.5
- RNase: None Detected
- Sterility: Passes test
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Color | Clear, Colorless |
| Quantity | 4 L |
| Grade | OmniPur, Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.