Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ PBS Tablets

Catalog No. 6501200TAB Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Pk./200 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-012-00TAB 1 Pk./200 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-012-00TAB Supplier MilliporeSigma™ Supplier No. 6501200TAB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Makes 1X solution when dissolved in 100mL distilled or deionized water from each tablet.

  • Molecular Biology Grade
  • DNase: None Detected
  • pH (1 tablet/100mL): 7.3-7.5
  • Protease: None Detected
  • RNase: None Detected
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color White
pH 7.3 to 7.5
Quantity 1 Pk./200 Tablets
Solution Type Buffer
Grade Molecular Biology, OmniPur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.