Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™, Protein Molecular Weight Marker, Wide Range

Catalog No. 7331500UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-315-00UL 500 μL/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73-315-00UL Supplier MilliporeSigma™ Supplier No. 7331500UL

For use in molecular biology applications.

  • Composed of 8 proteins
  • MW Range: 14.0 to 212 kd
  • Composition: Lysozyme 14.4 kd; Trypsin Inhibitor 20 kd; Carbonic Anhydrase 29 kd; Ovalbumin 35 kd; BSA 66 kd; Phosphorylase B 97 kd; β-Galactosidase 116 kd; Myosin 212 kd
  • Purity (8 distinct bs): Passes test
  • CAS: Glycerin: 56-81-5, Dithiothreitol - 3483-12-3, Water: 7732-18-5, Lysozyme: 12650-88-3
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, Colorless
Form Liquid
Quantity 500 μL/Pk.
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.