Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Sodium Dodecyl Sulfate Solution, 20%

Catalog No. 79911L Shop All MilliporeSigma Products
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
79-911-L 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-911-L Supplier MilliporeSigma™ Supplier No. 79911L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in molecular biology applications.

  • A260 (3%, water): ≤0.1 AU
  • A280 (3%, water): ≤0.1 AU
  • Assay (C-12 content): ≥99%
  • Conductivity: 21900 to 25500μmhos
  • DNAse: none detected
  • pH at 25°C: 5.0 to 8.0
  • Phosphate (PO4): ≤0.0005%
  • RNase: none detected
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.