Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ TBE Buffer, 10X

Catalog No. 88204L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-204-L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-204-L Supplier MilliporeSigma™ Supplier No. 88204L
Add to Cart
Add to Cart

Suitable for a wide range of molecular biology applications. Each liter of 10X TBE buffer yields 10 liters of 1X TBE buffer.

Final concentration of 1X TBE buffer: 0.0089 M Tris, 0.089 M Borate, 0.002 M EDTA. Molecular biology grade.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.