Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Orai1/CRAC Blocker, Cpd5J-4

Catalog No. 5306470001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-064-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-064-70001 Supplier MilliporeSigma™ Supplier No. 5306470001
Add to Cart
Add to Cart

A non-toxic thiourea derivative that acts as a blocker of Orai1, the pore subunit of Ca2+-release-activated Ca2+ (CRAC) in T

A non-toxic thiourea derivative that acts as a blocker of Orai1, the pore subunit of Ca2+-release-activated Ca2+ (CRAC) in T cells. Exhibits a strong blocking effect on store-operated Ca2+ entry in HeLa-O+S cells and in Th17 cells. Preferentially reduces Th17 differentiation and ameliorates Th17-mediated autoimmune disease. Blocks the infiltration of T cells into the CNS and reduces the number of IL-17 producing cells. Also shown to reduce the expression of retinoic acid receptor-related orphan receptors a (RORa) and RORgt. Reduces the symptoms and delays the onset of experimental autoimmune encephalomyelitis (EAE) in myelin oligodendrocyte glycoproteins 35-55 (MOG35-55) immunized mice (2mg/kg, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins Orail1
Molecular Formula C16H12N2O3S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.