Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ornithine Decarboxylase Inhibitor, POB

Catalog No. 49798510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-798-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-798-510MG Supplier MilliporeSigma™ Supplier No. 49798510MG
Add to Cart
Add to Cart

The Ornithine Decarboxylase Inhibitor, POB controls the biological activity of Ornithine Decarboxylase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Formulation Supplied as a hydrochloride salt.
Description 95% by HPLC
Molecular Formula C19H31N3O6
Formula Weight 397.5g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.