Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Oxidized ATP (oATP)

Catalog No. 5057580001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-575-80001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-575-80001 Supplier MilliporeSigma™ Supplier No. 5057580001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 2',3'-dialdehyde derivative of ATP that is commonly used to affinity label nucleotide sites in enzymes

A 2′,3′-dialdehyde derivative of ATP that is commonly used to affinity label nucleotide sites in enzymes. Acts as a specific, irreversible antagonist of P2Z/P2X7 purinergic receptors in J774 mouse macrophages. Does not affect P2γ receptors. Inhibits inflammatory pain in arthritic rat model by blocking ATP action on P2X7 receptor in nerve terminals. Also shown to irreversibly block ATP-induced Ca2+ influx in lymphocytes (˜100μM). O-ATP-treated mice display better preservation and lower rate of rejection of pancreatic islet grafts with reduced Th1 transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Product Type Antagonist
Target Proteins P2X7
Molecular Formula C10H14N5O13P3
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.