Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p-APMSF, Hydrochloride

Catalog No. 1782815MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-828-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-828-15MG Supplier MilliporeSigma™ Supplier No. 1782815MG
Add to Cart
Add to Cart

Alternative to DFP and PMSF.

Alternative to DFP and PMSF. An irreversible inhibitor of trypsin-like serine proteases. Inhibits trypsin and thrombin in equimolar concentration. Acts on C1r, C1s, Factor Xa, and plasmin, requiring a 5- to 10-fold excess for complete irreversible inhibition. Inhibitory activity is approximately 1000-fold greater than that of PMSF.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins trypsin-like serine proteases
CAS 74938-88-8
Formula Weight 252.7g/mol
Solubility H2O (100 mM).
Content And Storage Protect from moisture
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.