Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ P-Glycoprotein Inhibitor, C-4

Catalog No. 51730110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-730-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-730-110MG Supplier MilliporeSigma™ Supplier No. 51730110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The P-Glycoprotein Inhibitor, C-4 controls the biological activity of P-Glycoprotein. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins P-Glycoprotein
Molecular Formula C23H18ClNO4
Formula Weight 407.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.