Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p300/CBP Inhibitor VI, L002

Catalog No. 5052980001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-529-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-529-80001 Supplier MilliporeSigma™ Supplier No. 5052980001
Add to Cart
Add to Cart

A cell-permeable phenylsulfonyloxyiminocyclohexadienone compound that acts as a potent, selective, and reversible inhibitor of p300 histone acetyl transferase (IC50

A cell-permeable phenylsulfonyloxyiminocyclohexadienone compound that acts as a potent, selective, and reversible inhibitor of p300 histone acetyl transferase (IC50 = 1.98μM). Shown to occupy Ac-CoA binding pocket of p300, and less potent to CBP, PCAF, GCN5 and GNAT. Suppresses Histone H3 and H4 acetylation in triple negative breast cancer cell line MDAMB-468 as well as in HCT116 cell line and blocks p300-mediated STAT3 phosphorylation in pancreatic cancer cell line MIA Paca-2. Does neither inhibit the activities of histone deacetylases 1, 6, and 11 nor 8-diverse histone methyltransferases tested (DOT1, EZH1, G9a, PRMT1, SETD2, SET7-9, SMYD2, and SUV39H2). Reported to suppress tumor growth in mice bearing tumor xenografts of MDAMB-468 cell line (at 500μg/100μl, i.p. twice a week).
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 99% by HPLC
Target Proteins p300 HAT
Molecular Formula C15H15NO5S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.