Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p38 MAP Kinase Inhibitor VII, SD-169

Catalog No. 50615810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-615-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-615-810MG Supplier MilliporeSigma™ Supplier No. 50615810MG
Add to Cart
Add to Cart

The p38 MAP Kinase Inhibitor VII, SD-169, also referenced under CAS 1670-87-7, controls the biological activity of p38 MAP Kinase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 1670-87-7
Molecular Formula C9H8N2O
Formula Weight 160.2g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.