Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p38 MAP Kinase Inhibitor X, BIRB 796

Catalog No. 50617210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-617-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-617-210MG Supplier MilliporeSigma™ Supplier No. 50617210MG
Add to Cart
Add to Cart

The p38 MAP Kinase Inhibitor X, BIRB 796, also referenced under CAS 285983-48-4, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
CAS 285983-48-4
Molecular Formula C31H37N5O3
Formula Weight 527.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.