Learn More
Description
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
Specifications
| Color | Yellow |
| Description | 98% by HPLC |
| Target Proteins | p38a MAPK |
| Molecular Formula | C21H18/N4·2HCl |
| Solubility | H2O or DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 10 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.